BBL™ Yeast Fermentation Broth, BD Diagnostics
Supplier: BD Diagnostics
BBL™
297301
90004-824PK 75.36 USD
90004-824
BBL™ Yeast Fermentation Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Yeast fermentation media for the isolation of tubercle fungi.
  • With sucrose or maltose and Durham's tube
  • Sterile
  • Ready-to-use
The product includes a surface area of 5.4 cm for cultivation.