Anti-IL-23 (p19) Rat Monoclonal Antibody [clone: G23-8]

Supplier: BioXCell
BE0313-50MG BE0313-25MG BE0313-100MG BE0313-100MG BE0313-50MG BE0313-25MG
76556-046EA 4425.05 USD
76556-046 103778-356 76554-516 103778-358 76556-580 103778-360
Anti-IL-23 (p19) Rat Monoclonal Antibody [clone: G23-8]
Antibodies
Type: Primary
Antigen: IL-23 (p19)
Clonality: Monoclonal
Clone: G23-8
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR