Remotely monitor temperature by broadcasting readings to any Apple or Android smartphone or device with Bluetooth® 4.0 or better utilizing the free THERMSmart app.
- Monitor critical environments without a physical connection
- Maximum signal distance is approximately 40 meters
- Alarm sounds to alert user if temperature falls outside of the set parameters
- Alarm and timer settings savable for reuse
- LCD screen for on-unit monitoring
- App can trigger indicator light for unit identification
- Indicators include active alarm state and low battery
- Individually serialized
Verification model provides accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample.
Certifications: CE marked and RoHS compliant. Includes Certificate of Compliance indicating accuracy for 4 temperature points.
Delivery information: Batteries, multi-language (English, French, German, Italian, Portuguese and Spanish) instructions and countdown timer function are included.