Brij® 35 solution 10%, MegaPure™ detergent, sterile-filtered

Synonyms: Brij® L23Tricosaethylene glycol mono-n-dodecyl etherPolyoxyethylene(23)lauryl ether

B1184-50 B1184-25 B1184-100
76336-208EA 141.69 USD
76336-208 76336-206 76336-210
Brij® 35 solution 10%, MegaPure™ detergent, sterile-filtered
Brij® 35

The detergent Brij™ 35 is a nonionic polyoxyethylene surfactant frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications. The MegaPure® detergent solutions are carefully prepared to remove any aldehydes and peroxides. Can be used in cell culture applications.


  • A nonionic detergent

Formula: C₅₈H₁₁₈O₂₄
MW: 1199.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00080891
CAS Number: 9002-92-0
https://us.vwr.com/stibo/web/std.lang.all/35/82/27363582.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR