Portable Vein Finder, Springside Scientific
Supplier: Springside Scientific
PL800
76339-790EA 4268.5 USD
76339-790
Portable Vein Finder, Springside Scientific
Photodocumentation Systems
Primarily used to assist hospital laboratory staff locate difficult to see subcutaneous vessels during injections, IVs, and blood draws.
  • Portable device requires only simple assembly
  • Perfect for pediatric and dermatological applications
  • Long battery life, completely rechargeable
  • Light and portable
  • One-year limited warranty
The Portable Vein Finder provides clear and concise imaging with no harmful visible light, using Windows® based proprietary software. True 3D imaging enables the user to easily see the central point of the vessel, as well as a clear image of the needle tip itself.
This professional medical image processing system features a high resolution large screen, dual window mode display, with blood vessels clearly visible and displayed more accurately.
Certifications: FDA approved.