Isopropyl acetate ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Acetic acid isopropyl ester
45960-1L
45960-250ML
BJ45960-1LEA
158.36
USD
BJ45960-1L
BJ45960-250ML
Isopropyl acetate ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Isopropyl acetate
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Beilstein Number: 1740761
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 85 °C Density: 0.8662 g/cm³ (25 °C) Flash Pt: 2.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008877 CAS Number: 108-21-4 EINECS: 203-561-1 UN: 1220 ADR: 3,II |
Specification Test Results
assay (GC) | Min. 99 % |
measurement uncertainty | effective % |
identity (NMR) | complying |
refractive index (n 25/D) | 1.38 - 1.39 |
water (Karl Fischer) | Max. 0.5 % |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Autoignition Temperature | 894 °F |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...