Natural Square HDPE Bottle, with One-Touch Pump, Menda
Supplier: Menda
35396
89126-952EA 40.18 USD
89126-952
Natural Square HDPE Bottle, with One-Touch Pump, Menda
Bottles Plastic Bottles
The 4 oz. white translucent HDPE square bottle with one-touch pump is suitable for use with many solvents.
  • Imprint easily distinguishes bottle contents
  • Height: 3.375"
  • Dispenses approximately 0.2 cc of liquid with each pump
Bottle is imprinted with non-smearing, non-running ink, with the words 'isopropanol (IPA)'. Easily distinguish bottle contents due to exterior imprints. The square shape provides stability and saves storage space.