Giemsa's stain modified solution Azure blend, HARLECO®, Sigma-Aldrich®

Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

620-75 620-75
CA15204-146CS 2471.96 USD
CA15204-146 15204-146
Giemsa's stain modified solution Azure blend, HARLECO®, Sigma-Aldrich®
Giemsa's stain

Giemsa Stain Solution Modified Azure Blend HARLECO® Polychromatic stain used primarily with Wright Stain to stain blood cells and bone marrow specimens.


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use


Polychromatic stain used primarily with Wright Stain to stain blood cells and bone marrow specimens. Has various other uses in microbiology and histology staining.
Dilute with 50 volumes of distilled water or buffer immediately before use.

Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR