Giemsa's stain Azure B, HARLECO®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

619-71 619-71
15204-142EA 217.82 USD
CA15204-142 15204-142
Giemsa's stain Azure B, HARLECO®, Sigma-Aldrich®
Giemsa's stain

HARLECO® Polychromatic stain used alone or in combination with Wright Stain for staining of blood parasites


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use


Polychromatic stain used as a stand alone stain or in combination with Wright Stain for staining of blood parasites such as malaria and rickettsia.

Flash Pt: 11.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR