Hemacolor® Rapid staining of blood smear solution II, HARLECO®, Sigma-Aldrich®
Supplier: MilliporeSigma
65044B-85 65044B-85
15204-172EA 313.57 USD
15204-172 CA15204-172
Hemacolor® Rapid staining of blood smear solution II, HARLECO®, Sigma-Aldrich®
Hemacolor® Rapid staining of blood smear
HARLECO® Red color reagent for fast staining in hematology and cytology. Replacement for Red color reagent of the Hemacolor® Stain Set.
  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use
IVDLogo