Periodic acid solution 0.5%

Supplier: Leica Biosystems

Synonyms: Orthoperiodic acidortho-Periodic acid

Surgipath®
38016SS4A
75810-398EA 121.74 USD
75810-398
Periodic acid solution 0.5%
Periodic acid

Component of the Periodic Acid and Schiff Special Stain Kit


  • Differentiating solution


PAS staining is mainly used for staining structures containing a high proportion of carbohydrates such as glycogen, glycoproteins, proteoglycans typically found in connective tissues, mucus and basement membranes.

Formula: H₅IO₆
MW: 227.94 g/mol
MDL Number: MFCD00011344
CAS Number: 10450-60-9
https://us.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR