Shandon™ EA-65 stain for cytology
Supplier: Epredia
Synonyms:
EA 36
6766004
6766003
89370-820CS
91.44
USD
89370-820
89370-822
Shandon™ EA-65 stain for cytology
Papanicolaou's solution EA 65
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.