ProCulture® Complete Spinner Flask with Shur-Loc® Closure, Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
Supplier: SP Industries
BP-3600-114 BP-3600-110 BP-3600-116 BP-3600-112 BP-3600-108
76335-242EA 1421.38 USD
76335-242 76335-244 76335-246 76335-248 76335-240
ProCulture® Complete Spinner Flask with Shur-Loc® Closure, Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
For cultures that require a controlled environment, Shur-Loc® center neck caps have a unique compression O-ring design that provides a positive visible seal.
  • Flasks have internal baffles for enhanced agitation
  • Sidearm openings large enough for 50 ml pipettes
  • Flask base has central bump to prevent material from accumulating at the bottom center
  • Entire assembly made of FDA approved materials
  • Cap made from Polybutylene terephthalate (PBT), red rubber, Ultem®
  • Flask is type 1, class A borosilicate glass construction
  • All components are autoclavable
The impeller eliminates dead spots and extends above the media surface to increase aeration. It is made from PTFE, silicone, polysulfone, borosilicate glass.