MERC6039X
6039X-75 6039X-75
15204-260EA 177.86 USD
15204-260 CA15204-260
MERC6039X
HARLECO® Diluted solution of carbol fuchsin used as a counterstain in various staining modifications of the Gram Stain. Recommended for Hucker and Stirling methods.
  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use