Filter for Fine Particles, SCS, Desco Industries
Supplier: Desco Industries
SV-SPF1
89204-682EA 1.99 USD
89204-682
Filter for Fine Particles, SCS, Desco Industries
Filters Gas Filters
A thinner, more concentrated weave (HEPA media) for trapping extremely fine particles.
  • Intended to provide a dry environment for electronic devices that will be re-flow soldered
  • Efficiency: 99.97%
  • Size range: 0.3 to 0.5 µm
  • CFM: 40 cubic feet per minute
Typical applications include use with color laser printers, color copiers, and highly sensitive equipment. This pleated filter is encased in plastic, making it durable and easy to dispose.