Dri-Shield® Moisture Barrier Bags, 3000 Series, SCS, Desco Industries
Supplier: Desco Industries
D301030 D301020 D301618
89024-372PK 134.83 USD
89024-372 89024-370 89024-374
Dri-Shield® Moisture Barrier Bags, 3000 Series, SCS, Desco Industries
Bags Storage Bags
This foil moisture barrier bag provides protection for ESD sensitive and moisture sensitive electronics, typically SMT (Surface Mount Technology) items. Best practice is to use with Desiccant and a Humidity Indicator Card. A nylon layer helps to strengthen the bag. The bags are heat sealable and suitable for vacuum packaging. They are printed with ESD protective and moisture sensitive warning symbols, and a lot code for traceability.
  • 0.006" (6 mil) thick bag constructed from layers of nylon, foil, and polyethylene
  • </= 0.0003 Grams Moisture Vapor Transmission Rate (MVTR); designed for dry packing of moisture sensitive devices and to protect contents from damage due to ESD events or EMI
  • Nylon reinforcement layer is an ideal choice for packaging products with sharp corners
  • >16 lb puncture strength average; provides physical protection for contents
  • Dissipative inner and outer surfaces Rs 1×104 to < 1×1011 ohms per ANSI/ESD STM11.11; allows bag to be grounded when placed on a grounded surface or handled by a grounded operator
  • Lead-free RoHS 2, REACH and Conflict Minerals compliant
  • <10 nJ Energy Penetration Tested per ANSI/ESD STM11.31; protects contents from ESD charges and discharges
The metal layer provides discharge shielding and minimizes the penetration of electric fields and electrostatic discharges.
Certifications: Designed for moisture and electrostatic discharge (ESD) sensitive items (such as SMT). Meets ANSI/ESD S20.20, ANSI/ESD S541, ANSI/ESD S11.4 Level 1, and IPC/JEDEC J-STD-033.
Strengthens bag structure to maintain performance per ASTM F392 flex testing and meets the ANSI/ESD S541 requirement as a barrier to current flow.
Meets the requirement for discharge shielding per Static Control Bag ANSI/ESD S11.4 Level 1.
Meets the requirements of ANSI/ESD S541 for quality control purposes.