Sliding Microtomes, SM2010 R, Leica
Supplier: Leica
1492010RUEL 1492010RUSN 1492010RDSN
76001-008EA 26165.79 USD
76001-008 76001-012 76001-010
Sliding Microtomes, SM2010 R, Leica
Microtomes and Microtome Assemblies Microtomes
The stable sliding microtome has a totally enclosed micrometer feeding system with an ergonomically positioned object head close to the user. The smooth running sledge can be locked in 11 positions by using the easily accessible sledge brake.
  • Quick release system
  • Large volume antistatic section waste tray
  • Extended blade life
  • Ergonomically positioned object head
  • Totally enclosed micrometer feeding systems prevents debris from entering the roller bearings
With smooth movement of the sledge, precision specimen orientation, and emphasis on safety and ergonomics, the SM2010 R produces high quality sections for routine histopathology (paraffin).
Quick release system for specimen clamps allows operator to quickly and easily change from one clamp to another, resulting in improved workflow. Large volume antistatic section waste tray with area for tools allows collection of large volume of waste and ease of cleaning due to the antistatic surface, results in improved workflow and efficiency. Blade life is extended by a simple and safe lateral displacement function, resulting in cost-savings for the laboratory. Blade holder is recommended for soft tissue and biopsies.
Configuration 1 with universal cassette clamp and disposable blade holder SE for low profile blades.
Configuration 2 with universal cassette clamp and knife holder SN for reusable knives or blade rails and stable instrument construction.
Basic configuration with dry ice tray and knife holder SN and stable instrument construction.
Certifications: CE approved.