L(+)-Valine
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(S)-(+)-Valine
, Valine
, H-Val-OH
VA120-100GM
VA120-25GM
VA120-1KG
75873-554EA
603
USD
75873-554
75873-556
75873-552
L(+)-Valine
L(+)-Valine
L-Valine, Crystal, also known as 2-amino-3-methylbutanoic acid, is a branched-chain amino acid and is classified as non-polar; it can substitute for the hydrophilic amino acid glutamic acid in hemoglobin in sickle-cell disease.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064220 CAS Number: 72-18-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...