Undecanophenone

Supplier: SPECTRUM CHEMICAL MFG CORP

U1312-25GM U1312-100GM
75873-540EA 328.36 USD
75873-540 75873-282
Undecanophenone
Undecanophenone
Formula: C₁₇H₂₆O
Storage Temperature: Ambient
MDL Number: MFCD00051548
CAS Number: 4433-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR