L-Threonine

Synonyms: (2S,3R)-(-)-Threonine(2S,3R)-2-Amino-3-hydroxybutyric acidH-Thr-OH

TH150-1KG TH150-25GM TH150-100GM
75873-520EA 609.9 USD
75873-520 75873-522 75873-518
L-Threonine
L-Threonine

L-Threonine, Crystal, also known as 2-Amino-3-hydroxybutanoic acid, is an essential amino acid that is classified as polar. It bears a chiral side chain, as well as the amino acid, isoleucine. It is considered to be susceptible to posttranslational modifications.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR