5,6,7,8-Tetrahydro-2-naphthol

Supplier: SPECTRUM CHEMICAL MFG CORP

T1424-25GM T1424-100GM
75873-392EA 138 USD
75873-392 75873-390
5,6,7,8-Tetrahydro-2-naphthol
5,6,7,8-Tetrahydro-2-naphthol
Formula: C₁₀H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00001738
CAS Number: 1125-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR