1-Octanesulfonic acid sodium salt, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium 1-octansulfonateSodium octane-1-sulphonate

S2748-25GM S2748-100GM S2748-500GM
75873-334EA 312 USD
75873-334 75873-336 75873-338
1-Octanesulfonic acid sodium salt, Reagent Grade
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
https://us.vwr.com/stibo/web/std.lang.all/09/50/2300950.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR