Sodium 2,4-dinitrobenzenesulfonate, hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2,4-Dinitrobenzenesulphonic acid sodium salt

S1572-100GM S1572-25GM
75873-320EA 97.58 USD
75873-320 75873-318
Sodium 2,4-dinitrobenzenesulfonate, hydrate
Sodium 2,4-dinitrobenzenesulphonate
Formula: C₆H₃N₂NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00007471
CAS Number: 885-62-1
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR