Pyronin G

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Pyronine Y3,6-Bis(dimethylamino)xanthylium chloridePyronin Y

P1467-500GMBL P1467-1GM P1467-5GM P1467-25GM
75872-446EA 192 USD
75872-446 75872-444 75872-442 75873-160
Pyronin G
Pyronine G

Pyronin Y | Non-graded Pyronin Y supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₇H₁₉ClN₂O
MW: 302.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR