Polyvinylpyrrolidone cross-linked

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: PolyvinylpolypyrrolidoneN-Vinyl-2-Pyrrolidone

P1347-50GM P1347-500GM P1347-100GM
75873-146EA 92 USD
75873-146 75873-142 75873-144
Polyvinylpyrrolidone cross-linked
Polyvinylpyrrolidone cross-linked

Polyvinylpyrrolidone, Cross-linked, also known as called polyvidone or povidone, is a water-soluble polymer. It has been used as an adhesive, emulsifier, thickening agent and in molecular biology as a blocking agent during Southern blot analysis as a component of Denhardt's buffer.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₉NO
MW: 111.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 25249-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR