2-Naphthylamine

Supplier: SPECTRUM CHEMICAL MFG CORP

N2003-5KGBL N2003-1GM
75873-088EA 432668.08 USD
75873-088 75873-086
2-Naphthylamine
2-Naphthylamine

2-Naphthylamine, Powder, also known as β-Naphthylamine, is an aromatic amine and human carcinogen which can be used to make azo dyes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₉N
MW: 143.19 g/mol
Density: 1.061
Storage Temperature: Ambient
MDL Number: MFCD00004112
CAS Number: 91-59-8
UN: 1650
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR