3-Nitroaniline hydrochloride

Supplier: SPECTRUM CHEMICAL MFG CORP

N1514-25GM N1514-100GM
75873-054EA 316 USD
75873-054 75873-056
3-Nitroaniline hydrochloride
3-Nitroaniline hydrochloride
Formula: C₆H₇ClN₂O₂
MW: 174.5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050693
CAS Number: 33240-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR