Methyl tiglate

Supplier: SPECTRUM CHEMICAL MFG CORP

M1486-100ML M1486-25ML
75872-920EA 91.15 USD
75872-920 75872-918
Methyl tiglate
Methyl tiglate
Formula: C₆H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00016654
CAS Number: 6622-76-0
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR