3-(Hydroxymethyl)indole

Synonyms: Indole-3-methanol3-Indolcarbinol3-IndolmethanolIndole-3-carbinol(1H-Indol-3-yl)methanol(Indole-3-yl)methanol3-Indolemethanol

I2190-25GM I2190-1KGBL I2190-5GM I2190-100GM I2190-500GM
75872-810EA 1494.97 USD
75872-810 75872-808 75872-806 75872-804 75872-812
3-(Hydroxymethyl)indole
3-(Hydroxymethyl)indole

Indole-3-carbinol is most prevalent in cruciferous vegetables like cabbage, cauliflower, broccoli, collard greens, Brussels sprouts and kale. It has been studied for its possible anticarcinogenic, antioxidant, and anti-atherogenic effects.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₉NO
MW: 147.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005632
CAS Number: 700-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR