4-Isopropylaniline

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: p-Cumidine4-Aminocumene

I1272-100ML
75872-740EA 156.58 USD
75872-740
4-Isopropylaniline
4-Isopropylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007900
CAS Number: 99-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR