5-Hydroxy-DL-tryptophan

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: DL-5-Hydroxytryptophan

H2726-1GM H2726-5GM H2726-250MG H2726-100MG
75872-694EA 260.77 USD InStock
75872-694 75872-698 75872-696 75872-692
5-Hydroxy-DL-tryptophan
5-Hydroxy-DL-tryptophan
Formula: C₁₁H₁₂N₂O₃
MW: 220,23 g/mol
Melting Pt: 301 °C
Density: 1.405 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005651
CAS Number: 56-69-9
EINECS: 200-284-8
UN: 2811
ADR: 6.1,III
Merck Index: 13,04873

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR