4'-Hydroxyheptanophenone

Supplier: SPECTRUM CHEMICAL MFG CORP

H1518-100GM H1518-25GM
75872-654EA 139 USD
75872-654 75872-652
4'-Hydroxyheptanophenone
4'-Hydroxyheptanophenone
Formula: C₁₃H₁₈O₂
MW: 206.2800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191523
CAS Number: 14392-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR