Guanosine 5'-triphosphate trisodium salt
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
GTP trisodium salt
, (((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid, trisodium salt
, (((2R,3S,4R,5R)-5-(2-Amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)triphosphoric acid trisodium salt
Guanosine 5'-Triphosphate, Sodium Salt is a purine nucleoside triphosphate which plays a role as an activator of substrates in metabolic reactions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₁₃N₅Na₃O₁₄P₃ MW: 588.94 g/mol Storage Temperature: Low Temperature Freezer (–40°C) |
MDL Number:
MFCD00077781 CAS Number: 36051-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...