4-Ethylpyridine

Supplier: SPECTRUM CHEMICAL MFG CORP

E2111-25ML E2111-100ML
75872-484EA 67.63 USD
75872-484 75872-482
4-Ethylpyridine
4-Ethylpyridine
Formula: C₇H₉N
MW: 107.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006450
CAS Number: 536-75-4
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR