Ethyl-2-oxocyclohexanecarboxylate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethyl 2-oxocyclohexanecarboxylate

E1476-100ML E1476-25ML
75872-466EA 102.24 USD
75872-466 75872-464
Ethyl-2-oxocyclohexanecarboxylate
Ethyl 2-oxocyclohexanecarboxylate
Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001631
CAS Number: 1655-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR