Ethyl diethoxyacetate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Diethoxyacetic ethyl ester

E1444-25ML E1444-100ML
75872-396EA 92 USD
75872-396 75872-394
Ethyl diethoxyacetate
Ethyl diethoxyacetate
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009231
CAS Number: 6065-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR