cis-2,6-Dimethylpiperazine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R,6S)-2,6-Dimethylpiperazine

D1580-100GM D1580-25GM
75872-308EA 104 USD
75872-308 75872-306
cis-2,6-Dimethylpiperazine
cis-2,6-Dimethylpiperazine
Formula: C₆H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD07772435
CAS Number: 21655-48-1
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR