Clorsulon USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Amino-6-(1,2,2-trichlorovinyl)benzene-1,3-disulphonamide4-Amino-6-(trichlorovinyl)-1,3-benzenedisulphonamide4-Amino-6-(trichlorovinyl)-1,3-benzenedisulfonamide4-Amino-6-(1,2,2-trichlorovinyl)benzene-1,3-disulfonamide

C4533-1KGBL C4533-5KGBL C4533-100GM
76014-364EA 8992 USD
76014-364 76014-360 76014-362
Clorsulon USP
Clorsulon

Clorsulon, USP is anthelmintic agent and antibiotic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₈H₈Cl₃N₃O₄S₂
MW: 380.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072006
CAS Number: 60200-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR