Choline chloride USP dietary supplement
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-hydroxy-N,N,N-trimethylethanaminium chloride
C2058-2.5KG
C2058-500GM
C2058-12KG
76014-346EA
165
USD
76014-346
76014-324
76014-344
Choline chloride USP dietary supplement
Choline chloride
Choline Chloride, USP Dietary Supplement.
All Spectrum Chemical USP Dietary Supplement grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
HOH₂CCH₂N(CH₃)₃Cl MW: 139.62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011721 CAS Number: 67-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...