Benzaldehyde NF (DEA List I Chemical)

Supplier: SPECTRUM CHEMICAL MFG CORP

BE128-500MLGL BE128-200LTBL BE128-20LTMT BE128-4LTGL
76014-258EA 227.5 USD InStock
76014-258 76014-252 76014-254 76014-256
Benzaldehyde NF (DEA List I Chemical)
Benzaldehyde

Benzaldehyde (DEA List I Chemical), NF is used as a solvent in the preparation compound benzaldehyde elixirs. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₅CHO
MW: 106,12 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: -26 °C
Density: 1,043
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9,III
Merck Index: 13,01057
https://us.vwr.com/stibo/web/std.lang.all/60/95/746095.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR