Benzhydrylamine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aminodiphenylmethaneα-PhenylbenzylamineDiphenylmethanamine

B2577-100GM B2577-25GM
75871-836EA 392 USD
75871-836 75871-838
Benzhydrylamine
Benzhydrylamine
Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Density: 1.063
Storage Temperature: Ambient
MDL Number: MFCD00008059
CAS Number: 91-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR