Bis(2-ethylhexyl)phosphate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Di-(2-ethylhexyl)phosphoric acidBis(2-ethylhexyl) hydrogen phosphate

B2216-100GM B2216-25GM
75872-048EA 43.24 USD
75872-048 75872-046
Bis(2-ethylhexyl)phosphate
Bis(2-ethylhexyl) phosphate

Bis(2-ethylhexyl) Hydrogen Phosphate, also known as DEHA or Diisooctyl adipate, is a plasticizer used as a component of aircraft lubricants and sometimes as a functional hydraulic fluid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₅O₄P
MW: 322.43 g/mol
Density: 0.965
Storage Temperature: Ambient
MDL Number: MFCD00009492
CAS Number: 298-07-7
UN: 1902
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR