(Bromomethyl)cyclohexane

Supplier: SPECTRUM CHEMICAL MFG CORP

B1500-100GM B1500-25GM
75871-824EA 91.13 USD
75871-824 75871-822
(Bromomethyl)cyclohexane
(Bromomethyl)cyclohexane
Formula: C₇H₁₃Br
MW: 177.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001509
CAS Number: 2550-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR