β-Butyrolactone

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 1,3-ButyrolactoneButan-3-olide4-Methyl-2-oxetanonebeta-Methylpropiolactone4-Methyloxetan-2-onebeta-Butyrolactone

B1492-100ML B1492-25ML
75871-988EA 342 USD
75871-988 75871-990
β-Butyrolactone
β-Butyrolactone
Formula: C₄H₆O₂
MW: 86.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005170
CAS Number: 3068-88-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR