(±)-2-Bromoisovaleric acid

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Bromo-3-methylbutyricacid2-Bromo-3-methylbutyric acid

B1442-25GM B1442-100GM
75871-940EA 284.66 USD
75871-940 75871-942
(±)-2-Bromoisovaleric acid
(±)-2-Bromoisovaleric acid
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004210
CAS Number: 565-74-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR