4-(4-Nitrophenylazo)resorcinol

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Azo VioletMagneson I4-((4-Nitrophenyl)diazenyl)benzene-1,3-diolAzoviolet

AZ115-5GM AZ115-5KGBL AZ115-25GM AZ115-100GM
75871-912EA 56.09 USD
75871-912 75871-914 75871-908 75871-910
4-(4-Nitrophenylazo)resorcinol
4-(4-Nitrophenylazo)resorcinol

Azo Violet, also known as azoviolet or Magneson, is used as a dye and pH indicator.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₉N₃O₄
MW: 259.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007310
CAS Number: 74-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR