6-Amino-1-naphthalenesulfonic acid

Supplier: SPECTRUM CHEMICAL MFG CORP

A3986-100GM
75871-878EA 200.39 USD
75871-878
6-Amino-1-naphthalenesulfonic acid
6-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00035724
CAS Number: 81-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR