Aminoguanidine hydrochloride
Supplier: SPECTRUM CHEMICAL MFG CORP
A3814-40KGBL
A3814-100GM
A3814-25GM
76014-186EA
12194
USD
76014-186
75871-864
75871-866
Aminoguanidine hydrochloride
Aminoguanidine hydrochloride
Aminoguanidine Hydrochloride, also known as Pimagedine or Guanyl hydrazine, is a nitric oxide and diamine oxidase synthase inhibitor which can be used for energetic material applications.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
CH₇ClN₄ MW: 110.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00039074 CAS Number: 1937-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...