Bis(2-methoxyethyl)adipate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Adipic acid bis(2-methoxyethyl) ester

A2914-100ML A2914-25ML
75871-850EA 261 USD
75871-850 75871-852
Bis(2-methoxyethyl)adipate
Bis(2-methoxyethyl) adipate
Formula: C₁₂H₂₂O₆
MW: 262.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041905
CAS Number: 106-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR