1-Amino-4-methylpiperazine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-Methylpiperazin-1-amine

A2075-100ML A2075-25ML
75871-728EA 263 USD
75871-728 75871-730
1-Amino-4-methylpiperazine
1-Amino-4-methylpiperazine
Formula: C₅H₁₃N₃
MW: 115.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006154
CAS Number: 6928-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR